Azure B (azure I), brown powder C.I. 52010
Leverantör: MP Biomedicals
Synonyms: C.I. 52010, N,N,N′-Trimethylthionin, Azure B, Trimethylthionine chloride, 3-(Dimethylamino)-7-(methylamino)phenothiazin-5-ium chloride, Methylene Azur B
0215041705 0215041701
ICNA0215041705EA 1250 SEK
ICNA0215041705 ICNA0215041701
Azure B (azure I), brown powder C.I. 52010
Azure B (azure I)
Azure B is used for differentiating cellular RNA (purplish) and DNA (blue-green) in plant tissues. Useful as a stain for Negri Bodies. Can be used in the Romanowsky-type stains for blood and blood protozoa.
Soluble in water (50 mg/ml) or ethanol (8-10 mg/ml); insoluble in diethyl ether.
Formel: C₁₅H₁₆ClN₃S
MW: 305,83 g/mol
Smältpunkt: 205…210 °C (dec., lit.)
Storage Temperature: Omgivande
MDL Number: MFCD00011935
CAS nummer: 531-55-5
EINECS: 208-511-2