5-Bromo-2,4-dimethoxypyrimidine ≥98.0%
Leverantör: TCI EUROPE
Varning
B1920-1G B1920-25G B1920-5G
TCIAB1920-1GEA 590 SEK
TCIAB1920-1G TCIAB1920-25G TCIAB1920-5G
5-Bromo-2,4-dimethoxypyrimidine ≥98.0%
5-Bromo-2,4-dimethoxypyrimidine
Formel: C₆H₇BrN₂O₂
MW: 219,0400 g/mol
Kokpunkt: 125°C
MDL Number: MFCD00038016
CAS nummer: 56686-16-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c0696e68f571de12270f1c61021440fd30d8637&vid=4c0696e68f571de12270f1c61021440fd30d8637&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 62.0 to 65.0 deg-C