(-)-Di-p-toluoyl-L-tartaric acid ≥98%
Leverantör: Thermo Scientific
Synonyms: Di-p-toluoyl-L-tartaric acid, (-)-O,O'-Di-p-toluoyl-L-tartaric acid, (2R,3R)-(-)-Di-O-4-Toluoyl-L-Tartaric Acid
B22972.14 B22972.22
B22972.14EA 2350 SEK
B22972.14 B22972.22
(-)-Di-p-toluoyl-L-tartaric acid ≥98%
(-)-Di-p-toluoyl-L-tartaric acid
Purity: 98% CAS No.: 32634-66-5 Molecular Formula: C20H18O8 Formula Weight: 386.36 Melting Point: 166-171° Beilstein Reference: 2022481
Formel: C₂₀H₁₈O₈
MW: 386,36 g/mol
Smältpunkt: 166…171 °C
Storage Temperature: Omgivande
MDL Number: MFCD00064440
CAS nummer: 32634-66-5
EINECS: 251-131-7