Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine) 97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: N1,N1'-(Butane-1,4-diyl)bis(propane-1,3-diamine), Spermine
L19562.03 L19562.06
L19562.03EA 1050 SEK
L19562.03 L19562.06
Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine) 97%
Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine)
Purity: 97% CAS No.: 71-44-3 Molecular Formula: C10H26N4 Formula Weight: 202.35 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 28-30° Boiling Point: 150°/5mm Beilstein Reference: 1751791 Merck Reference: 14,8743 UN No.: 3259 MDL No.: MFCD00008215
Formel: C₁₀H₂₆N₄
Kokpunkt: 150 °C (5 mmHg)
Smältpunkt: 28…30 °C
Flampunkt: >110 °C (230 °F)
MDL Number: MFCD00008215
CAS nummer: 71-44-3
EINECS: 200-754-2
ADR: 8,III
Merck Index: 14,8743