Lithium ethoxide 1 mol/l i etanol, AcroSeal®
Leverantör: Thermo Fisher Scientific
Fara
380171000 380178000
380171000.EA 1575 SEK
380171000. 380178000.
Lithium ethoxide 1 mol/l i etanol, AcroSeal®
Lithium ethoxide
Formel: C₂H₅LiO
Densitet: 0,82 g/cm³
Flampunkt: 9 °C
Storage Temperature: Omgivande
MDL Number: MFCD00050493
CAS nummer: 2388-07-0
UN: 3274
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Colorless to orange Solution
Strength 0.90 to 1.10 M