4,6-Dihydroxy-2-methylpyrimidine ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Methylpyrimidine-4,6-diol, 2-Methyl-4,6-pyrimidinediol
A10535.09 A10535.18
A10535.09EA 1050 SEK
A10535.09 A10535.18
4,6-Dihydroxy-2-methylpyrimidine ≥99%
4,6-Dihydroxy-2-methylpyrimidine
Purity: 99% CAS No.: 40497-30-1 Molecular Formula: C5H6N2O2 Formula Weight: 126.12 Melting Point: 360° Beilstein Reference: 115646
Formel: C₅H₆N₂O₂
MW: 126,12 g/mol
Smältpunkt: >360 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006104
CAS nummer: 40497-30-1
EINECS: 254-941-9