Tiglic acid 98%
Leverantör: Thermo Scientific
Fara
Synonyms: (E)-2-Methylbut-2-enoic acid, trans-2,3-Dimethylacrylic acid
138970250 138971000 138975000
ACRO138970250EA 595 SEK
ACRO138970250 ACRO138971000 ACRO138975000
Tiglic acid 98%
Tiglic acid
Formel: C₅H₈O₂
MW: 100,12 g/mol
Smältpunkt: 61…65 °C
Densitet: 0,96 g/cm³
Flampunkt: 95 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00066864
CAS nummer: 80-59-1
EINECS: 201-295-0
UN: 3261
ADR: 8,III
Order Now
Specifikation testresultat
Appearance White to beige Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 61 to 65 °C
Titration with NaOH ≥97.5 %