N-Methylisobutylamine ≥98.0%
Leverantör: TCI
|
Fara
|
M0691-25ML
M0691-100ML
TCIAM0691-25MLEA
1150
SEK
TCIAM0691-25ML
TCIAM0691-100ML
N-Methylisobutylamine ≥98.0%
N-Methylisobutylamine
Formel:
C₅H₁₃N Kokpunkt: 78°C |
MDL Number:
MFCD00015043 CAS nummer: 625-43-4 EINECS: 210-893-0 UN: 2733 ADR: 3,II |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...