N-Methylisobutylamine ≥98%

Leverantör: Thermo Fisher Scientific
Fara

42947.03 42947.06
42947.03EA 725 SEK
42947.03 42947.06
N-Methylisobutylamine ≥98%
N-Methylisobutylamine

Purity: 98+% CAS No.: 625-43-4 Molecular Formula: CH3NHCH2CH(CH3)2 Formula Weight: 87.16 Physical Form: Liquid Density: 0.73 Boiling Point: 78° UN No.: 2733 MDL No.: MFCD00015043

Formel: C₅H₁₃N
Kokpunkt: 78 °C
Densitet: 0,73
Storage Temperature: Omgivande
MDL Number: MFCD00015043
CAS nummer: 625-43-4
EINECS: 210-893-0
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR