Automatic saccharimeters, ADS420
Leverantör: BELLINGHAM & STANLEY
37-20-61-181 37-22-61-181 37-22-61-182 37-20-61-182 37-20-61-191 37-22-61-191 37-21-61-181 37-21-61-182 37-21-61-191
BELL37-20-61-181EA 94050 SEK
BELL37-20-61-181 635-0765 635-0766 BELL37-20-61-182 BELL37-20-61-191 635-0767 635-0761 635-0762 635-0763
Automatic saccharimeters, ADS420
Polarimetrar
Automatic saccharimeter designed for sugar purity in laboratories and tare houses.
  • PC software for 'purity' calculation
  • LED light source that requires no maintenance
  • Suitable for the measurement of samples with low transmittance
A single temperature sensor provides the measurement for sugar compensation whilst quartz compensation facilitates accurate verification and calibration using a quartz control plate by accessing the span facility in the setup menu.
These saccharimeters can measure dark samples of up to 3 OD, for optimum performance it is ideal to remove colour and suspended solids to provide a clear solution or one of less than 2,0 OD.
Certifikat: ICUMSA