Cocarboxylase, EMPROVE® api suitable for use as an active pharmaceutical ingredient, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Synonyms: Thiamine pyrophosphate chloride, Aneurinepyrophosphoric acid, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-((hydroxy(phosphonooxy)phosphoryl)oxy)ethyl)-4-methylthiazol-3-ium chloride, Cocarboxylase chloride
5.00112.5000 5.00112.1000 5.00112.0100
5.00112.5000EA 0 SEK
5.00112.5000 5.00112.1000 5.00112.0100
Cocarboxylase, EMPROVE® api suitable for use as an active pharmaceutical ingredient, SAFC®
Cocarboxylase
Formel: C₁₂H₁₉ClN₄O₇P₂S
MW: 460.77 g/mol
MDL Number: MFCD00038740
CAS nummer: 154-87-0
EINECS: 205-836-1