AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride), Sigma-Aldrich®
Leverantör: Merck
Varning
Synonyms: 4-(2-Aminoethyl)benzenesulphonyl fluoride hydrochloride, AEBSF hydrochloride
MQ100
1.24839.0100 1.24839.0500
1.24839.0100EA 920 SEK
1.24839.0100 1.24839.0500
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride), Sigma-Aldrich®
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)
Formel: C₈H₁₁ClFNO₂S
MW: 239.69 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00132962
CAS nummer: 30827-99-7
UN: 3261