N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-12, SB-12), Millipore®
Leverantör: Merck Millipore Calbiochem
|
Fara
|
Synonyms:
sulphobetaine-12, 3-(Dodecyldimethylammonio)propane-1-sulphonate, N-Dodecyl-n,n-dimethyl-3-ammonio-1-propanesulphonate
693015-25
693015-5
693015-1KG
693015-25EA
12850
SEK
693015-25
693015-5
693015-1KG
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-12, SB-12), Millipore®
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-12, SB-12)
Formel:
C₁₇H₃₇NO₃S MW: 335,55 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00036909 CAS nummer: 14933-08-5 EINECS: 239-002-3 UN: 3261 ADR: 8,II |