N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-10, SB-10), Millipore®
Leverantör: Merck Millipore Calbiochem
Varning
Synonyms: 3-[Dec-1-yl(dimethyl)ammonio]propane-1-sulphonate, 3-(Decyldimethylazaniumyl)propane-1-sulphonate
693021-100 693021-25 693021-5
693021-100EA 42850 SEK
693021-100 693021-25 693021-5
N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-10, SB-10), Millipore®
N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-10, SB-10)
Formel: C₁₅H₃₃NO₃S
MW: 307,4924 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00036908
CAS nummer: 15163-36-7