3-Fluoro-4-methoxybenzyl bromide ≥98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 4-(Bromomethyl)-2-fluoro-1-methoxybenzene
H26116.MD H26116.03
H26116.MDEA 1075 SEK
H26116.MD H26116.03
3-Fluoro-4-methoxybenzyl bromide ≥98%
3-Fluoro-4-methoxybenzyl bromide
Purity: 98% CAS No.: 331-61-3 Molecular Formula: C8H8BrFO Formula Weight: 219.05 Odour: Pungent Melting Point: 44-46° UN No.: 3261 MDL No.: MFCD00671768
Formel: C₈H₈BrFO
MW: 219.05 g/mol
Smältpunkt: 44…46 °C
Storage Temperature: Omgivande
MDL Number: MFCD00671768
CAS nummer: 331-61-3
UN: 3261
ADR: 8 II