Cyclohexyl isocyanide 99%
Leverantör: Thermo Fisher Scientific
Varning
158400010 158400050 158400250
ACRO158400010EA 850 SEK
ACRO158400010 ACRO158400050 ACRO158400250
Cyclohexyl isocyanide 99%
Cyclohexyl isocyanide
Formel: C₇H₁₁N
Kokpunkt: 173…176 °C (760 mmHg)
Densitet: 0.89 g/cm³
Flampunkt: 77 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00003839
CAS nummer: 931-53-3
EINECS: 213-238-7
UN: 2810
ADR: 6.1 III
Order Now
Specifikation testresultat
Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4490 to 1.4520 (20°C, 589 nm)
GC ≥98.5 %
UV at 500 nm A: ≤0.02
pH 8.5 to 9.6 (10 % in water)
Titration with HClO4 ≥98.5 %