2-Mercapto-5-methylbenzimidazole
Leverantör: Thermo Scientific
Synonyms:
1,3-Dihydro-5-methyl-2H-benzimidazole-2-thione, 2-Mercapto-5-methyl-1H-benzimidazole
SB00799EE
MAYBSB00799EEEA
0
SEK
MAYBSB00799EE
2-Mercapto-5-methylbenzimidazole
2-Mercapto-5-methylbenzimidazole
Formel:
C₈H₈N₂S MW: 164,23 g/mol Kokpunkt: 349 °C (1013 hPa) Smältpunkt: 220 °C |
MDL Number:
MFCD00010617 CAS nummer: 27231-36-3 EINECS: 248-350-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...