4-Methylbenzyl mercaptan

Leverantör: Thermo Scientific

Synonyms: p-Xylene-α-thiol

TL00457EA
MAYBTL00457EAEA 0 SEK
MAYBTL00457EA
4-Methylbenzyl mercaptan
4-Methylbenzyl mercaptan
Formel: C₈H₁₀S
MW: 138,23 g/mol
Kokpunkt: 116…118 °C (35 mmHg)
Densitet: 1,034 g/cm³ (23 °C)
MDL Number: MFCD00039652
CAS nummer: 4498-99-1
EINECS: 224-796-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR