Orotic acid monohydrate 98%
Leverantör: Thermo Scientific
Varning
Synonyms: Orotic acid hydrate, 2,6-Dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate
129571000 129575000
ACRO129571000EA 1175 SEK
ACRO129571000 ACRO129575000
Orotic acid monohydrate 98%
Orotic acid monohydrate
Formel: C₅H₄N₂O₄·1H₂O
MW: 156,10 g/mol (anhy)
Smältpunkt: 334 °C
Storage Temperature: Omgivande
MDL Number: MFCD00149398
CAS nummer: 50887-69-9
EINECS: 200-619-8
Order Now
Specifikation testresultat
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤12.0 % (1 g, 130°C, 3 hrs)
Titration with NaOH ≥97.5 %