Cyanamide 50% (w/w) i vattenlösning stabilised
Leverantör: Thermo Fisher Scientific
|
Fara
|
L20448.0B
L20448.22
L20448.30
L20448.0BEA
3700
SEK
L20448.0B
L20448.22
L20448.30
Cyanamide 50% (w/w) i vattenlösning stabilised
Cyanamide
CAS No.: 420-04-2
Molecular Formula: CH2N2
Formula Weight: 42.04
Storage Temperature: 2°C to 8°C
Flash Point: None
Refractive Index: 1.3980
Odour: Odourless
pH: 4.2
Beilstein Reference: 1732569
Merck Reference: 14,2684
UN No.: 2922
MDL No.: MFCD00007572
Formel:
CH₂N₂ MW: 42.04 g/mol Densitet: 1.06 Storage Temperature: Kylskåp |
MDL Number:
MFCD00007572 CAS nummer: 420-04-2 EINECS: 206-992-3 UN: 2922 ADR: 8 III Merck Index: 14,2684 |