β-Cyclodextrine ≥98%, Calbiochem®, Millipore®
Leverantör: Merck Millipore Calbiochem
Synonyms: Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex
239760-25 239760-500
239760-25EA 1625 SEK
239760-25 239760-500
β-Cyclodextrine ≥98%, Calbiochem®, Millipore®
β-Cyclodextrine
Formel: C₄₂H₇₀O₃₅
MW: 1134,98 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00078139
CAS nummer: 7585-39-9
EINECS: 231-493-2