Methyl-3-aminothiophene-2-carboxylate ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: Methyl 3-amino-2-thiophenecarboxylate
A12802.14 A12802.06 A12802.22
A12802.14EA 2900 SEK
A12802.14 A12802.06 A12802.22
Methyl-3-aminothiophene-2-carboxylate ≥98%
Methyl-3-aminothiophene-2-carboxylate
Purity: 98+% CAS No.: 22288-78-4 Molecular Formula: C6H7NO2S Formula Weight: 157.19 Melting Point: 63-67° Boiling Point: 100-102°/0.1mm MDL No.: MFCD00009765
Formel: C₆H₇NO₂S
MW: 157,19 g/mol
Kokpunkt: 100…102 °C (0,1 mmHg)
Smältpunkt: 63…67 °C
Storage Temperature: Omgivande
MDL Number: MFCD00009765
CAS nummer: 22288-78-4
EINECS: 244-895-8