2-(Phenylthio)nicotinic acid
Leverantör: Thermo Scientific
BTB08722DA
BTB08722EA
MAYBBTB08722DAEA
0
SEK
MAYBBTB08722DA
MAYBBTB08722EA
2-(Phenylthio)nicotinic acid
Formel:
C₁₂H₉NO₂S |
MDL Number:
MFCD00052110 CAS nummer: 35620-72-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...