2-(Methylthio)nicotinic acid ≥98%
Leverantör: Thermo Fisher Scientific
|
Varning
|
L09920.09
L09920.04
L09920.09EA
815
SEK
L09920.09
L09920.04
2-(Methylthio)nicotinic acid ≥98%
2-(Methylthio)nicotinic acid
Purity: 98+%
CAS No.: 74470-23-8
Molecular Formula: C7H7NO2S
Formula Weight: 169.20
Melting Point: 215-218°
Beilstein Reference: 4391503
Formel:
C₇H₇NO₂S Smältpunkt: 215…218 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00010101 CAS nummer: 74470-23-8 |