(R)-(-)-Prolinol ≥99.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: D-Prolinol, (2R)-pyrrolidin-2-ylmethanol, D(-)-Prolinol
P1274-1G P1274-25G P1274-5G
TCIAP1274-1GEA 215 SEK
TCIAP1274-1G TCIAP1274-25G TCIAP1274-5G
(R)-(-)-Prolinol ≥99.0% (by GC, titration analysis)
(R)-(-)-Prolinol
Formel: C₅H₁₁NO
MW: 101,1 g/mol
Flampunkt: 86°C
MDL Number: MFCD00064321
CAS nummer: 68832-13-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=938d9fff00f697d59399c47c90e486b25eddc74c&vid=938d9fff00f697d59399c47c90e486b25eddc74c&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 99.0 %
Specific rotation [a]20/D -29.0 to -33.0 deg (C=1, Toluene)