2-Bromo-4-(trifluoromethyl)phenylacetonitrile ≥98%
Leverantör: Thermo Scientific
Fara
H53518.03 H53518.06
H53518.03EA 1425 SEK
H53518.03 H53518.06
2-Bromo-4-(trifluoromethyl)phenylacetonitrile ≥98%
2-Bromo-4-(trifluoromethyl)phenylacetonitrile
Purity: 98% CAS No.: 474024-36-7 Molecular Formula: C9H5BrF3N Formula Weight: 264.04 Refractive Index: 1.5020 Boiling Point: 265-266° MDL No.: MFCD03094308
Formel: C₉H₅BrF₃N
MW: 264,04 g/mol
Kokpunkt: 265…266 °C
Densitet: 1,626
Storage Temperature: Omgivande
MDL Number: MFCD03094308
CAS nummer: 474024-36-7