3-(Trimethoxysilyl)propyl methacrylate 98%
Leverantör: Thermo Scientific
Varning
Synonyms: γ-methacryloxypropyl-trimethoxysilane, 3-(Methacryloyloxy)propyltrimethoxysilane
216550050 216551000 216555000
ACRO216550050EA 455 SEK
ACRO216550050 ACRO216551000 ACRO216555000
3-(Trimethoxysilyl)propyl methacrylate 98%
3-(Trimethoxysilyl)propyl methacrylate
Formel: C₁₀H₂₀O₅Si
MW: 248,35 g/mol
Kokpunkt: 253 °C (1017hPa)
Densitet: 1,04 g/cm³
Flampunkt: 100 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008593
CAS nummer: 2530-85-0
EINECS: 219-785-8
Order Now
Specifikation testresultat
Appearance Clear liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4300 to 1.4320 (20°C, 589 nm)
GC ≥97.5 %
Color scale ≤70 APHA