3,6-Dithia-1,8-octanediol ≥97%
Leverantör: Thermo Scientific
Synonyms: 1,8-Dihydroxy-3,6-Dithiaoctane, 2,2-(Ethane-1,2-diylbis(sulphanediyl))diethanol
A17654.09 A17654.18 A17654.30
A17654.09EA 505 SEK
A17654.09 A17654.18 A17654.30
3,6-Dithia-1,8-octanediol ≥97%
3,6-Dithia-1,8-octanediol
Purity: 97% CAS No.: 5244-34-8 Molecular Formula: HOCH2CH2SCH2CH2SCH2CH2OH Formula Weight: 182.31 Melting Point: 63-67° Boiling Point: 170°/0.8mm MDL No.: MFCD00002911
Formel: C₆H₁₄O₂S₂
MW: 182,31 g/mol
Kokpunkt: 170 °C (0,8 mmHg)
Smältpunkt: 63…67 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002911
CAS nummer: 5244-34-8
EINECS: 226-045-8