Glycine methyl ester hydrochloride (H-Gly-OMe.HCl) ≥99%
Leverantör: Thermo Scientific
Synonyms: Methyl glycinate hydrochloride, H-Gly-OMe.HCl, Glycine Methyl Ester HCl
A14083.0E A14083.22 A14083.36
A14083.0EEA 10750 SEK
A14083.0E A14083.22 A14083.36
Glycine methyl ester hydrochloride (H-Gly-OMe.HCl) ≥99%
Glycine methyl ester hydrochloride (H-Gly-OMe.HCl)
Purity: 99% CAS No.: 5680-79-5 Molecular Formula: H2NCH2CO2CH3.HCl Formula Weight: 125.56 Melting Point: ca 175° dec. Beilstein Reference: 3593644
Formel: C₃H₈ClNO₂
MW: 125,56 g/mol
Smältpunkt: ∼ 175 °C (dec.)
Densitet: 1,000
Storage Temperature: Omgivande
MDL Number: MFCD00012870
CAS nummer: 5680-79-5
EINECS: 227-139-1