Trimethylurea

Leverantör: Thermo Fisher Scientific

16475.03 16475.06 16475.14
16475.03EA 885 SEK
16475.03 16475.06 16475.14
Trimethylurea
Trimethylurea

CAS No.: 632-14-4 Molecular Formula: (CH3)2NC(O)NH(CH3) Formula Weight: 102.14 Physical Form: Solid/semi-solid mass Sensitivity: Hygroscopic Odour: Characteristic MDL No.: MFCD00014868

Formel: C₄H₁₀N₂O
Smältpunkt: 74…75 °C
Storage Temperature: Omgivande
MDL Number: MFCD00014868
CAS nummer: 632-14-4
EINECS: 211-169-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR