2-Amino-2-cyanoacetamide 97+%
Leverantör: Thermo Fisher Scientific
Fara
382760050 382760250
ACRO382760050EA 1125 SEK
ACRO382760050 ACRO382760250
2-Amino-2-cyanoacetamide 97+%
2-Amino-2-cyanoacetamide
Formel: C₃H₅N₃O
Smältpunkt: 120…124 °C
Storage Temperature: Omgivande
MDL Number: MFCD00040532
CAS nummer: 6719-21-7
EINECS: 229-771-3
UN: 2928
ADR: 6.1,II
Order Now
Specifikation testresultat
Appearance Beige to tan Powder
Infrared spectrum Conforms
Melting point 120 to 124 °C
Titration with HClO4 ≥97.0 %