Methyl (methoxymethylene)acetate 95%
Leverantör: Thermo Scientific
Varning
Synonyms: Methyl 3-methoxyacrylate
330460250
ACRO330460250EA 590 SEK
ACRO330460250
Methyl (methoxymethylene)acetate 95%
Methyl (methoxymethylene)acetate
Formel: C₅H₈O₃
MW: 116,1152 g/mol
Kokpunkt: 169 °C (760 mmHg)
Smältpunkt: 3…4 °C
Densitet: 1,07 g/cm³
Flampunkt: 65 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00128206
CAS nummer: 34846-90-7
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4490 to 1.4510 (20°C, 589 nm)
GC ≥94 %