(R)-(+)-2-Pyrrolidinone-5-carboxylic acid ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: D-Pyr-OH, (R)-(+)-2-Pyrrolidone-5-carboxylic acid
B24678.22 B24678.06 B24678.14
B24678.22EA 14750 SEK
B24678.22 B24678.06 B24678.14
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid ≥98%
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid
Purity: 98+% CAS No.: 4042-36-8 Molecular Formula: C5H7NO3 Formula Weight: 129.12 Melting Point: 158-160° Beilstein Reference: 82133
Formel: C₅H₇NO₃
MW: 129,12 g/mol
Smältpunkt: 158…160 °C
Storage Temperature: Omgivande
MDL Number: MFCD00066212
CAS nummer: 4042-36-8
EINECS: 223-735-0