Orotic acid, vattenfri 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 2,6-Dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid
B25349.36 B25349.22
B25349.36EA 6350 SEK
B25349.36 B25349.22
Orotic acid, vattenfri 97%
Orotic acid
Purity: 97% CAS No.: 65-86-1 Molecular Formula: C5H4N2O4 Formula Weight: 156.10 Melting Point: 344-347° MDL No.: MFCD00006027
Formel: C₅H₄N₂O₄
MW: 156,10 g/mol
Smältpunkt: 344…347 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006027
CAS nummer: 65-86-1
EINECS: 200-619-8