2-Dimethylaminoethyl methacrylate 97% Stabilised
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: DMA
Stabilization: Hydrochinonmonomethylether
L13507.AE L13507.AP
L13507.AEEA 745 SEK
L13507.AE L13507.AP
2-Dimethylaminoethyl methacrylate 97% Stabilised
2-Dimethylaminoethyl methacrylate
Stabilized with ca 0.2% 4-methoxyphenol
Purity: 97% CAS No.: 2867-47-2 Molecular Formula: C8H15NO2 Formula Weight: 157.21 Storage Temperature: 2°C to 8°C Flash Point: 70°(158°F) Refractive Index: 1.4400 Melting Point: ca -30° Boiling Point: 186-188° Beilstein Reference: 1757048 UN No.: 2522 MDL No.: MFCD00008589
Formel: C₈H₁₅NO₂
MW: 157.21 g/mol
Kokpunkt: 186…188 °C
Smältpunkt: ∼–30 °C
Densitet: 0.932
Flampunkt: 70 °C (158 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00008589
CAS nummer: 2867-47-2
EINECS: 220-688-8
UN: 2522
ADR: 6.1 II