2,5-Dichlorothiophene-3-carbonyl chloride

Leverantör: Thermo Scientific

Synonyms: 2,5-Dichlorothiophene-3-carbonylchloride

CD09458DA CD09458EA
MAYBCD09458DAEA 0 SEK
MAYBCD09458DA MAYBCD09458EA
2,5-Dichlorothiophene-3-carbonyl chloride
2,5-Dichlorothiophene-3-carbonyl chloride
Formel: C₅HCl₃OS
MW: 215.4848 g/mol
MDL Number: MFCD00041441
CAS nummer: 57248-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR