Diuron (N-(3,4-dichlorophenyl)-N',N'-dimethylurea) ≥97%
Leverantör: Thermo Scientific
Varning
L02986.18 L02986.30
L02986.18EA 990 SEK
L02986.18 L02986.30
Diuron (N-(3,4-dichlorophenyl)-N',N'-dimethylurea) ≥97%
Diuron (N-(3,4-dichlorophenyl)-N',N'-dimethylurea)
Purity: 97% CAS No.: 330-54-1 Molecular Formula: C9H10Cl2N2O Formula Weight: 233.10 Odour: Odourless Melting Point: 157-160° Boiling Point: 180-190° dec. Beilstein Reference: 2215168 Merck Reference: 14,3382 UN No.: 3077 MDL No.: MFCD00018136
Formel: C₉H₁₀Cl₂N₂O
MW: 233,10 g/mol
Kokpunkt: 180…190 °C (dec.)
Smältpunkt: 157…160 °C
Densitet: 1,48
Storage Temperature: Omgivande
MDL Number: MFCD00018136
CAS nummer: 330-54-1
EINECS: 206-354-4
ADR: 9,III
Merck Index: 14,3382