4,4''-Diamino-p-terphenyl 95%
Leverantör: Thermo Fisher Scientific
|
Varning
|
L05403.03
L05403.06
L05403.03EA
2125
SEK
L05403.03
L05403.06
4,4''-Diamino-p-terphenyl 95%
4,4''-Diamino-p-terphenyl
Purity: 95%
CAS No.: 3365-85-3
Molecular Formula: C18H16N2
Formula Weight: 260.34
Melting Point: 244-250°
Beilstein Reference: 2120295
Formel:
C₁₈H₁₆N₂ Smältpunkt: 244…250 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00051532 CAS nummer: 3365-85-3 |