2,5-Dibromo-3-methylthiophene ≥98%
Leverantör: Thermo Scientific
Varning
L02693.06 L02693.14
L02693.06EA 895 SEK
L02693.06 L02693.14
2,5-Dibromo-3-methylthiophene ≥98%
2,5-Dibromo-3-methylthiophene
Purity: 98% CAS No.: 13191-36-1 Molecular Formula: C5H4Br2S Formula Weight: 255.97 Refractive Index: 1.6130 Boiling Point: 226-230° MDL No.: MFCD00015470
Formel: C₅H₄Br₂S
MW: 255,97 g/mol
Kokpunkt: 226…230 °C
Densitet: 1,972
Storage Temperature: Omgivande
MDL Number: MFCD00015470
CAS nummer: 13191-36-1
EINECS: 236-147-4