Diethyl tetradecylphosphonate 98%
Leverantör: Thermo Fisher Scientific
B21426.06 B21426.14
B21426.06EA 1825 SEK
B21426.06 B21426.14
Diethyl tetradecylphosphonate 98%
Diethyl tetradecylphosphonate
Purity: 98% CAS No.: 5191-09-3 Molecular Formula: C18H39O3P Formula Weight: 334.47 Refractive Index: 1.4445 Melting Point: 8° Boiling Point: 199-201°/3mm MDL No.: MFCD00015597
Formel: C₁₈H₃₉O₃P
Kokpunkt: 199…201 °C (3 mmHg)
Smältpunkt: 8 °C
Densitet: 0,922
Storage Temperature: Omgivande
MDL Number: MFCD00015597
CAS nummer: 5191-09-3