(±)-1-Phenylethanol-[D9] (98% D) for NMR spectroscopy
Leverantör: Merck
Fara
Synonyms: (±)-1-Phenylethanol-[²H₉]
8.15057.0001 8.15057.0005
8.15057.0001EA 0 SEK
8.15057.0001 8.15057.0005
(±)-1-Phenylethanol-[D9] (98% D) for NMR spectroscopy
(±)-1-Phenylethanol-[D9]
Formel: C₆H₅CH(OH)CH₃
Storage Temperature: Omgivande
CAS nummer: 19547-01-4
UN: 2937
ADR: 6.1,III