4-tert-Butylcalix[6]arene 96%
Leverantör: Thermo Scientific
Varning
Synonyms: p-tert-Butylcalix[6]arene
L14473.03 L14473.06 L14473.14
L14473.03EA 625 SEK
L14473.03 L14473.06 L14473.14
4-tert-Butylcalix[6]arene 96%
4-tert-Butylcalix[6]arene
Purity: 96% CAS No.: 78092-53-2 Molecular Formula: C66H84O6 Formula Weight: 973.40 Melting Point: 377-381° Beilstein Reference: 2611918
Formel: C₆₆H₈₄O₆
MW: 973,39 g/mol
Smältpunkt: 377…381 °C
Storage Temperature: Omgivande
MDL Number: MFCD00075464
CAS nummer: 78092-53-2