Calix[4]arene 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 1,3,5,7(1,3)-Tetrabenzenacyclooctaphane-12,32,52,72-tetraol
290190010 290190050
ACRO290190010EA 1800 SEK
ACRO290190010 ACRO290190050
Calix[4]arene 98%
Calix[4]arene
Formel: C₂₈H₂₄O₄
MW: 424,5 g/mol
Smältpunkt: 315…318 °C
Storage Temperature: Omgivande
MDL Number: MFCD00233673
CAS nummer: 74568-07-3
Order Now
Specifikation testresultat
Appearance Off-white to light beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %