(Trifluoromethyl)trimethylsilane 0.5 M i THF
Leverantör: Thermo Scientific
Fara
Synonyms: trimethyl(trifluoromethyl)silane
368320050 368320250 368321000
ACRO368320050EA 435 SEK
ACRO368320050 ACRO368320250 ACRO368321000
(Trifluoromethyl)trimethylsilane 0.5 M i THF
(Trifluoromethyl)trimethylsilane
Formel: C₄H₉F₃Si
MW: 142,2 g/mol
Kokpunkt: 40 °C (760 mmHg)
Densitet: 0,895 g/cm³
Flampunkt: –20 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00145454
CAS nummer: 81290-20-2
UN: 1993
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless Solution
Strength 0.45 to 0.55 M