(±)-1,2,3,4-Tetrahydro-1-naphthylamine 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Tetralin-1-amine, 1,2,3,4-Tetrahydronaphthalen-1-amine
B25673.06 B25673.14
B25673.06EA 1100 SEK
B25673.06 B25673.14
(±)-1,2,3,4-Tetrahydro-1-naphthylamine 97%
(±)-1,2,3,4-Tetrahydro-1-naphthylamine
Purity: 97% CAS No.: 2217-40-5 Molecular Formula: C10H13N Formula Weight: 147.22 Flash Point: 110°(230°F) Refractive Index: 1.5640 Boiling Point: 246-247°/714mm MDL No.: MFCD00001740
Formel: C₁₀H₁₃N
MW: 147.22 g/mol
Kokpunkt: 246…247 °C (714 mmHg)
Densitet: 1.026
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00001740
CAS nummer: 2217-40-5
EINECS: 218-712-7