Hafnium diboride ≥99.5% (max. 2% Zr)
Leverantör: Thermo Scientific
Synonyms: Hafnium boride
43664.30 43664.09 43664.18
43664.30EA 28000 SEK
43664.30 43664.09 43664.18
Hafnium diboride ≥99.5% (max. 2% Zr)
Hafnium diboride
Purity: 99.5% CAS No.: 12007-23-7 Molecular Formula: B2Hf Formula Weight: 200.11 Physical Form: -325 Mesh Powder Density: 10.5 Melting Point: 3250° MDL No.: MFCD00016125
Formel: B₂Hf
MW: 200,11 g/mol
Smältpunkt: 3250 °C
Densitet: 10,5
Storage Temperature: Omgivande
MDL Number: MFCD00016125
CAS nummer: 12007-23-7
EINECS: 234-500-7