4-Hydroxy-2(5H)-furanone 95%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Tetronic acid, beta-Tetronic acid, 2,4(3H,5H)-Furandione, Tetrahydrofuran-2,4-dione
235260010 235260050
ACRO235260010EA 970 SEK
ACRO235260010 ACRO235260050
4-Hydroxy-2(5H)-furanone 95%
4-Hydroxy-2(5H)-furanone
Formel: C₄H₄O₃
MW: 100.07 g/mol
Smältpunkt: 135…137 °C
Storage Temperature: Omgivande
MDL Number: MFCD00052187
CAS nummer: 541-57-1
EINECS: 208-785-3
Order Now
Specifikation testresultat
Appearance Light yellow to beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥94 %