(1-Naphthoxy)acetic acid ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-(Naphthalen-1-yloxy)acetic acid
L01036.06 L01036.14
L01036.06EA 965 SEK
L01036.06 L01036.14
(1-Naphthoxy)acetic acid ≥98%
(1-Naphthoxy)acetic acid
Purity: 98+% CAS No.: 2976-75-2 Molecular Formula: C12H10O3 Formula Weight: 202.21 Melting Point: 193-197° Beilstein Reference: 1912165
Formel: C₁₂H₁₀O₃
MW: 202,21 g/mol
Smältpunkt: 193…197 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003927
CAS nummer: 2976-75-2
EINECS: 221-023-4