Potassium methyl malonate 98%
Leverantör: Thermo Scientific
Varning
Synonyms: Monomethyl monopotassium malonate, Monomethyl malonate potassium salt
269550250 269551000 269555000
ACRO269550250EA 805 SEK
ACRO269550250 ACRO269551000 ACRO269555000
Potassium methyl malonate 98%
Potassium methyl malonate
Formel: C₄H₅KO₄
MW: 156,18 g/mol
Smältpunkt: >200 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00014021
CAS nummer: 38330-80-2
EINECS: 253-886-8
Order Now
Specifikation testresultat
Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %