Triethylenetetramine 60%
Leverantör: Thermo Scientific
|
Fara
|
Synonyms:
N,N'-Bis(2-aminoethyl)-1,2-ethanediamine, Trientine, DEH 24, EPH 925
344070500
344075000
344070025
ACRO344070500EA
465
SEK
ACRO344070500
ACRO344075000
ACRO344070025
Triethylenetetramine 60%
Triethylenetetramine
Formel:
C₆H₁₈N₄ MW: 146,24 g/mol Kokpunkt: 280 °C (760 mmHg) Smältpunkt: –35 °C Densitet: 0,98 g/cm³ Flampunkt: 129 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00008169 CAS nummer: 112-24-3 EINECS: 203-950-6 UN: 2259 ADR: 8,II |
Specifikation testresultat
Appearance | Clear Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4960 to 1.5000 (20°C, 589 nm) |
GC | ≥55 % |
Color scale | ≤150 APHA |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...